Learn More
STEMCELL Technologies SmartDish™ - 5 Pack
Click to view available options
Quantité:
5/paquet
Description
- SmartDish™ plates enable accurate and reproducible hematopoietic colony counting by preventing the formation of a meniscus
- This allows for an even distribution of culture medium, resulting in a more uniform distribution of colonies throughout the entire 35 mm well
- The absence of the meniscus also reduces optical distortion so that colonies located at the edge can be more easily counted
- SmartDish™ (Catalog #27370) is the second generation of the original SmartDish™ plate (Catalog #27301)
- These new plates use an improved coating, which streamlines the manufacturing process while consistently improving the visibility of hematopoietic colonies in culture through elimination of the mensicus
- Current SmartDish™ customers can contact their local STEMCELL representative for more information
- SmartDish™ has been designed for both manual counting with STEMgrid™-6 as well as automated colony counting with a STEMvision™ instrument
Spécification
Spécification
| À utiliser avec (application) | Cell Culture; Colony Assay |
| Quantité | 5/paquet |