Learn More
STEMCELL Technologies SmartDish™ - 5 Pack
Code produit. 15761668
Encompass_Preferred
Click to view available options
Quantité:
5/paquet
Code produit. 15761668
Marque: STEMCELL Technologies 27370
Meniscus-free cultureware for more accurate counting of hematopoietic colonies in CFU assays
  • SmartDish™ plates enable accurate and reproducible hematopoietic colony counting by preventing the formation of a meniscus
  • This allows for an even distribution of culture medium, resulting in a more uniform distribution of colonies throughout the entire 35 mm well
  • The absence of the meniscus also reduces optical distortion so that colonies located at the edge can be more easily counted
  • SmartDish™ (Catalog #27370) is the second generation of the original SmartDish™ plate (Catalog #27301)
  • These new plates use an improved coating, which streamlines the manufacturing process while consistently improving the visibility of hematopoietic colonies in culture through elimination of the mensicus
  • Current SmartDish™ customers can contact their local STEMCELL representative for more information
  • SmartDish™ has been designed for both manual counting with STEMgrid™-6 as well as automated colony counting with a STEMvision™ instrument
Spécification
À utiliser avec (application) Cell Culture; Colony Assay
Quantité 5/paquet
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.