Learn More
Lenz Laborglasintrumente Sockets
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marque: Lenz Laborglasintrumente 01100134
Code nomenclature Nacres: NB.43
Informations supplémentaires : Poids : 0.10000kg
Description
- Conforme à la norme DIN 12249, formulaire E, en complément de la norme DIN
- Prises rodées coniques
- En tubulure DURAN™
Spécification
NS 34/35 | |
Verre Duran |