Learn More
DiscoverX Sodium L-lactate
Code produit. 17362529
Encompass
Click to view available options
:
10 mg
Code produit. 17362529
Marque: DiscoverX 921209
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement
Sodium L-lactate is an agonist for GPR81 (HCAR1). Provided as 10mg of dry powder.
Sodium L-lactate is a sodium salt of lactic acid and an agonist for GPR81 (HCAR1). It is produced from pyruvate during anaerobic glycolysis and in proliferatively active cells as a result of reaction with the enzyme lactate dehydrogenase. Used as a food additive, it acts as a preservative, acidity regulator, and bulking agent. Also used in cleaning supplies like shampoo and soap, and can be used to treat arrhythmias, as a pressor sympathomimetic, and to prevent and control metabolic acidosis. Provided as 10mg of dry powder.
Spécification
Quantité 10 mg