Learn More
Specially Denatured Alcohol (SDA) 2B-3 200 Proof, Greenfield
Click to view available options
Quantité:
4x1 Fluorinated Poly Gallon Case
55 Gallon
Conditionnement:
15.14L
208.20L
Spécification
Spécification
| Nom chimique ou matériau | Specially Denatured Alcohol (SDA) 2B-3 200 Proof |
| Point d’ébullition | 78.5 |
| Quantité | 4x1 Fluorinated Poly Gallon Case |
| Conditionnement | 4x1 Fluorinated Poly Gallon Case |