Learn More
Bolle Safety Spectrum Goggle
GREENER_CHOICE
Code produit 31810811
Change view
Click to view available options
Frame Type:
Indirect Vented
Indirect Vented with Foam
Sealed
Sealed with Foam
Teinte de la lentille:
Clear
Smoke
Modèle:
SPECTN10W
SPECTN11W
SPECTN12W
SPECTN13W
SPECTN14W
SPECTN15W
SPECTN20W
Conditionnement:
1 pièce
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Frame Type Teinte de la lentille Modèle unitSize
31810811 Sealed Clear SPECTN15W 1 pièce
31810813 Indirect Vented Smoke SPECTN20W 1 pièce
31810799 Indirect Vented Clear SPECTN14W 1 pièce
31810775 Sealed with Foam Clear SPECTN13W 1 pièce
31810781 Sealed Clear SPECTN12W 1 pièce
31810787 Indirect Vented with Foam Clear SPECTN11W 1 pièce
31810806 Indirect Vented Clear SPECTN10W 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Code produit 31810811 Fournisseur Bolle Safety Code fournisseur SPECTN15W
With its bi-material frame and adjustable strap, Spectrum offers superior protection and adaptability. Available in two sizes, Spectrum is a versatile product with different frame types and lens coatings to provide a solution for every company, regardless of their specific work environment. Its sporty design and optimized ergonomics also ensure greater acceptance for enhanced safety and productivity.
  • PC/TPR Co-injected frame for optimum comfort.
  • Multiple coatings and sizes available.
  • Soft gasket to fit most face structures.
  • Optimal panoramic vision.
TRUSTED_SUSTAINABILITY
Spécification
Revêtement de la lentille Platinum
Teinte de la lentille Clear
Type de produit Safety Goggles
Modèle SPECTN15W
Matériau Polycarbonate
Jetable Reusable
Lens Material Polycarbonate
Matériau de la monture Polycarbonate
Protection UV None
Frame Type Sealed
Autoclavable Not Autoclavable