Learn More
Invitrogen™ Human SRC N1 Recombinant Protein
Code produit. 10728323
Click to view available options
Quantité:
1 mg
Conditionnement:
1mg
Code produit. 10728323
Marque: Invitrogen™ PV4087
Recombinant Protein
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.
SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth. SRCN1 is a neuronal splice variant of SRC.
TRUSTED_SUSTAINABILITY
Spécification
Numéro d’adhésion P12931
Concentration See Label
À utiliser avec (application) Kinase Assay
Formule 50mM tris with 0.05% NP-40, 1mM DTT, 50% glycerol, 0.05mM EDTA, 100mM NaCl and no preservative; pH 7.5
Identification génétique (Entrez) 6714
Poids moléculaire 60.6 kDa
Nom Human SRC N1
Plage de pH 7.5
Méthode de purification Purified
Quantité 1 mg
Conditions de stockage -80°C, Avoid Freeze/Thaw Cycles
État réglementaire RUO
Alias de gène SRCN1
Nom usuel SRC N1
Symbole de gène(s) SRC
Famille des kinases SRC Kinase Family
Type de produit Protein
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Séquence Full length
Contenu et stockage -80°C, Avoid Freeze/Thaw Cycles
Groupe de kinases Tyrosine Kinases
Conditions d’expédition Dry Ice
Système d’expression Baculovirus
Forme protéique Full Length
Forme Liquid
Catégorie de recherche Drug Development