Learn More
Stainless Steel, AISI 316L, S31603, ARMI∣MBH
Code produit. 30002419
Click to view available options
Conditionnement:
1 pièce
Code produit. 30002419
Marque: ARMI/MBH IARM163EX
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Stainless Steel, AISI 316L, S31603, Ø38x3mm
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Lead/Zirconium/Boron/Antimony/Calcium/Aluminium/Oxygen/Arsenic/Tin/Carbon/Titanium/Phosphorus/Niobium/Tungsten/Nitrogen/Vanadium/Cobalt/Copper/Silicon/Manganese/Nickel/Chromium/Molybdenum
CAS 7439-89-6
Forme Disc
Concentration or Composition (by Analyte or Components) Lead (<0.0005) %/Zirconium (0.001) %/Boron (0.0019) %/Antimony (0.002) %/Calcium (0.002) %/Aluminium (0.0039) %/Oxygen (0.007) %/Arsenic (0.008) %/Tin (0.012) %/Carbon (0.014) %/Titanium (0.02) %/Phosphorus (0.028) %/Niobium (0.034) %/Tungsten (0.035) %/Nitrogen (0.061) %/Vanadium (0.063) %/Cobalt (0.115) %/Copper (0.47) %/Silicon (0.53) %/Manganese (1.48) %/Nickel (10.17) %/Chromium (16.74) %/Molybdenum (2.05) %