missing translation for 'onlineSavingsMsg'
Learn More

Temsirolimus, MedChemExpress

Code produit. 16477896
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantité:
5 mg
10 mg
25 mg
100 mg
This item is not returnable. View return policy

Product Code. 16477896

missing translation for 'mfr': Medchem Express HY50910

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model.

Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model.

Specifications

Nom chimique ou matériau Temsirolimus
Forme physique Powder
CAS 162635-04-3
Couleur Blanc
Quantité 100 mg
Notes de qualité de la pureté Research
Formule moléculaire C56H87NO16
À utiliser avec (application) Cancer-Kinase/protease
Synonyme CCI-779
Informations sur la solubilité DMSO : 250 mg/mL (242.65 mM; Need ultrasonic) ∣Ethanol : 200 mg/mL (194.12 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@H]4C[C@H]([C@H](OC(C(C)(CO)CO)=O)CC4)OC)C)=O)=O
Poids moléculaire (g/mol) 1030.29
Poids de la formule 1030.29
Pourcentage de pureté 98.0%
Qualité Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.