Learn More
Tenovin-3, MedChemExpress
Product Code. 16473636
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantité:
1 mg
5 mg
10 mg
25 mg
50 mg
This item is not returnable. View return policy
Product Code. 16473636
missing translation for 'mfr': Medchem Express HY19339
This item is not returnable. View return policy
MedChemExpress Tenovin-3 is a p53 activator.
Tenovin-3 is a p53 activator.
Specifications
Nom chimique ou matériau Tenovin-3
Forme physique Solid
CAS 1011301-27-1
Couleur Blanc
Quantité 25 mg
Notes de qualité de la pureté Research
Formule moléculaire C18H21N3OS
À utiliser avec (application) Cancer-programmed cell death
Informations sur la solubilité DMSO : ≥ 31 mg/mL (94.67 mM)
SMILES O=C(NC(NC1=CC=C(N)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2
Poids moléculaire (g/mol) 327.44
Poids de la formule 327.44
Pourcentage de pureté 99.86%
Qualité Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.