missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Terrific Broth, Powder
Click to view available options
Quantité:
500 g
1 kg
5 kg
Conditionnement:
1kg
500g
5kg
Description
Enriched media for maintenance and propogation of E.
Enriched media for maintenance and propogation of E. coli and plasmid growth. Produces higher cell densities resulting in greater yields of DNA or recombinant problem. Glycerol is used as a carbon source with this formulation instead of glucose as it will not ferment to acetic acid.
Ingredients:
Casein Digest Peptone: 12 g/l
Yeast Extract: 24 g/l
Dipotassium phosphate: 9.4 g/l
Monopotassium phosphate: 2.2 g/l
Specifications
Specifications
| Quantité | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction