missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TFPI Recombinant Protein

Code produit. 16141352
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16141352

Marque: Abnova™ H00007035P01.10ug

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Human TFPI full-length ORF recombinant protein with GST-tag at N-terminal

This gene encodes a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. The product of this gene inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop. The encoded protein is glycosylated and predominantly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been confirmed.

  • Molecular Weight: 53.35kDa
  • Preparation Method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer

Sequence: MIYTMKKVHALWASVCLLLNLAPAPLNADSEEDEEHTIITDTELPPLKLMHSFCAFKADDGPCKAIMKRFFFNIFTRQCEEFIYGGCEGNQNRFESLEECKKMCTRDNANRIIKTTLQQEKPDFCFLEEDPGICRGYITRYFYNNQTKQCERFKYGGCLGNMNNFETLEECKNICEDGPNGFQVDNYGT
QLNAVNNSLTPQSTKVPSLFVTKEGTNDGWKNAAHIYQVFLNAFCIHASMFFLGLDSISCLC

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spécification

Numéro d’adhésion AAH15514
À utiliser avec (application) Antibody Production, Protein Array, ELISA, Western Blot
Formule 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Identification génétique (Entrez) 7035
Poids moléculaire 53.35
Nom TFPI (Human) Recombinant Protein (P01)
Plage de pH 8
Méthode de préparation In vitro wheat germ expression system
Méthode de purification Glutathione Sepharose 4 Fast Flow
Test du contrôle qualité 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantité 10 μg
Source Wheat Germ (in vitro)
Immunogène MIYTMKKVHALWASVCLLLNLAPAPLNADSEEDEEHTIITDTELPPLKLMHSFCAFKADDGPCKAIMKRFFFNIFTRQCEEFIYGGCEGNQNRFESLEECKKMCTRDNANRIIKTTLQQEKPDFCFLEEDPGICRGYITRYFYNNQTKQCERFKYGGCLGNMNNFETLEECKNICEDGPNGFQVDNYGTQLNAVNNSLTPQSTKVPSLFVTKEGTNDGWKNAAHIYQVFLNAFCIHASMFFLGLDSISCLC
Conditions de stockage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Alias de gène EPI/LACI/TFI/TFPI1
Nom usuel TFPI
Symbole de gène(s) TFPI
Réactivité croisée Human
Espèces Wheat Germ (in vitro)
Recombinant Recombinant
Marqueur de protéine GST
Forme Solution
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis