Learn More
Thermo Scientific™ Flat PCR Cap Strips
Code produit. 11564794
Click to view available options
Quantité:
300
Conditionnement:
300 pièces
Code produit. 11564794
Marque: Thermo Scientific™ AB0603
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement
Barrettes de bouchons bombés pour applications PCR à utiliser avec les plaques PCR à 96 puits et les barrettes de tubes Thermo Scientific.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Thermo Scientific Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
• Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
• Large end tabs for strip removal (12 strips only)
Autoclavable Resistance:Successfully Tested
> DMSO (100%) Resistance:Successfully Tested
> Ethanol Resistance:Successfully Tested
> Gamma Irradiation Resistance:Successfully Tested
> Hazardous:No
>Isopropanol Resistance:Successfully Tested
> Long Term Storage:Successfully Tested
> PCR (including waterbath):Successfully Tested
> Peelable:Successfully Tested
> Pierceable:Not Recommended
> qPCR:Not Recommended
> Resealable:Successfully Tested
> Sealing Temperature Range:-20°C to +120°C
> Storage Condition:Ambient
> UV Irradiation Resistance:Successfully Tested
Certifications
Certifié exempt d’ADN humain, de RNase et de DNase.
TRUSTED_SUSTAINABILITY
Spécification
Certifications/Conformité Certified free from human DNA, RNases, and DNases
Quantité 300