Learn More
Thermo Scientific™ Hypersil GOLD™ C8 Javelin™ Guard Columns
Tous les produits Thermo Scientific Produits
Click to view available options
Diamètre (métrique) interne:
3 mm
Granulométrie:
5 μm
Conditionnement:
4 pièces
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
Features of Hypersil GOLD C8 Solid Phase
- Hypersil GOLD C8 columns provide similar selectivity to C18 columns but with reduced retention
- Lower hydrophobicity
- Faster separations
- Excellent peak shape
- High efficiency
- Outstanding sensitivity
Informations sur la commande
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
Spécification
Spécification
| Granulométrie | 5 μm |
| Longueur (métrique) | 10 mm |
| Diamètre (métrique) interne | 3 mm |
| Phase stationnaire | C8 |