Learn More
Cole-Parmer Thermoplastic Elastomer (TPE) Tubing
Tous les produits Cole-Parmer Produits
Click to view available options
Diamètre (impériale) interne:
0,0625 po
0,125 po
0,188 po
0,25 po
0,313 po
0,5 po
0,625 po
0,75 po
1/32 po.
1 po
3/32 po
3/8 in.
Diamètre (impériale) externe:
0,0938 po
0,25 po
0,313 po
0,375 po
0,438 po
0,5 po
0,625 po
0,75 po
0,878 po
1 in
1,25 po
1/8 po
3/16 po
5/32 po
7/32 po
9 à 16 po
Conditionnement:
15m
Description
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Spécification
Spécification
| Certifications/Conformité | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diamètre (impériale) interne | 3/32 po |
| Matériau | TPE |
| Diamètre (impériale) externe | 5/32 po |
| Longueur totale (impériale) | 50 ft. |
| Longueur totale (métrique) | 50 ft. |
| Méthode de stérilisation | EtO with no issues |
| Couleur | Clear |