Learn More
BOLA Tiles
Click to view available options
Dimensions (L x l):
300 x 2 mm
300 x 3 mm
300 x 4 mm
300 x 5 mm
300 x 6 mm
300 x 8 mm
300 x 10 mm
300 x 15 mm
Conditionnement:
1 pièce
Description
Standard sizes with different thicknesses.
Spécification
Spécification
| Matériau | PTFE |
| Absorbance | Non-absorbent |
| Dimensions (L x l) | 300 x 2 mm |
| Jetable | Single-use |