Learn More
Thermo Scientific Chemicals TCL grade 12 microns, gel de silice APS
Click to view available options
Quantité:
50 g
250 g
1 kg
5 kg
Conditionnement:
1kg
250g
50g
5kg
Description
TLC plate preparation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Granulométrie | 12 μm |
| Porosité | 60 Å |
| Qualité | TLC |
| pH | 6,5 à 7,5 |
| Quantité | 50 g |
| Aire de la surface (métrique) | 500 à 600 m2/g |
| Type | Gel de silice |
| Matériau | Silice |