missing translation for 'onlineSavingsMsg'
Learn More

Trifloxystrobin, MedChemExpress

Code produit. p-200101104
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantité:
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 16495816

missing translation for 'mfr': Medchem Express HY123230

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress Trifloxystrobin (CGA 279202) is a fungicide, with EC50s of 23.0 μg/L and 1.7 μg/L for Daphnia magna neonate and embryos, respectively, after treatment for 48 h.

Trifloxystrobin (CGA 279202) is a fungicide, with EC50s of 23.0 μg/L and 1.7 μg/L for Daphnia magna neonate and embryos, respectively, after treatment for 48 h.

Specifications

Nom chimique ou matériau Trifloxystrobin
Forme physique Solid
CAS 141517-21-7
Couleur Blanc
Quantité 100 mg
Notes de qualité de la pureté Research
Formule moléculaire C20H19F3N2O4
À utiliser avec (application) COVID-19-immunoregulation
Synonyme CGA 279202
Informations sur la solubilité DMSO : 125 mg/mL (306.09 mM; Need ultrasonic)
SMILES C/C(C1=CC(C(F)(F)F)=CC=C1)=N\OCC2=C(C=CC=C2)/C(C(OC)=O)=N\OC
Poids moléculaire (g/mol) 408.37
Poids de la formule 408.37
Pourcentage de pureté 99.68%
Qualité Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.