Learn More
DiscoverX Tyramine
Code produit. 17372539
Encompass
Click to view available options
Quantité:
10 mg
Code produit. 17372539
Marque: DiscoverX 921223
Tyramine is an agonist for TAAR1. Provided as 10mg of dry powder.
Tyramine is a naturally occuring monoamine derivative of tyrosine that acts as a catecholamine releasing agent and agonist of TAAR1. Has nonpsychoactive peripheral sympathomimetic effects, and can lead to a hypertensive crisis if foods rich in tyramine are ingested in conjunction with monoamine oxidase inhibitors. Occurs widely in foods as a result of fermentation or decay via tyrosine decarboxylation. Provided as 10mg of dry powder.
Spécification
Quantité 10 mg