missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- With bound neck and internal seams
- Available in sizes from Small to 3X
- Covered elastication at wrists
- Front snap closure for easy donning and doffing
- Not clean-processed and not sterilized
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splashes
- Breathable and exceptionally low linting
- Manufactured in a controlled environment
- Typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry and electronics industries as well as in other critical or controlled environments
- 30 per box, bulk packed
Specifications
Specifications
| Type de produit | Controlled Environments Lab Coat |
| Dimensions | X-Large |
| Matériau | Polyethylene |
| Couleur | Blanc |
| Type de bouchon | Snaps |
| Type de manchette | Élastique |
| Jetable | Single-use |
| Stérilité | Non stérile |
| Type de collier | Bound Neck |
| À utiliser avec (application) | Biotech, Pharmaceuticals, Electronics, Medical Device Manufacturing, Food Processing |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?