missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Umeclidinium bromide, MedChemExpress
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
Description
Umeclidinium bromide is a novel mAChR antagonist. The affinity (Ki) of Umeclidinium bromide for the cloned human M1-M5 mAChRs ranges from 0.05 to 0.16 nM.
Specifications
Specifications
| Nom chimique ou matériau | Umeclidinium bromide |
| Forme physique | Solid |
| CAS | 869113-09-7 |
| Couleur | Off-White |
| Quantité | 100 mg |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C29H34BrNO2 |
| À utiliser avec (application) | COVID-19-immunoregulation |
| Synonyme | GSK573719A |
| Informations sur la solubilité | DMSO : ≥ 34 mg/mL (66.86 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction