Learn More
Brady™ V4500 Barcode Scanner Kit
Code produit. p-200022864
Click to view available options
Comprend:
Lecteur de codes-barres V4500
Lecteur de codes-barres V4500, batterie V1200
Lecteur de codes-barres V4500, socle V1300, batterie V1200, adaptateur secteur, câble d'alimentation, câble USB de 1 m
Conditionnement:
1 kit
1 pièce
Règlementaire : Cet article est soumis à éco-contribution.
Code produit. 30057752
Marque: Brady™ 177233
This item is not returnable. View return policy
Rugged design. Seamless experience. Digitize data capture and automate your workflows with ease and accuracy with the V4500 Barcode Scanner.
  • High-speed, omnidirectional barcode scanning made simple.
  • Ideal for inventory management, kit verification, scan and print, and packout applications.
  • Proprietary Cortex Decoder technology allows scanner to easily read tiny barcodes as well as shiny and hard-to-read surfaces.
  • Automated keyboard device and dynamic user interface lets you go from setup to first scan in seconds.
  • Data parsing feature lets you sort and arrange data exactly how you want – start with a simple scan and easily connect to the cloud and your ERP system.
  • Withstands 6 ft. drops (MIL-STD-810G).
Specifications
Description V4500 Barcode Reader
Autonomie de la batterie Up to 75,000 scans per charge
Types de code-barre pris en charge BC412, Codabar, Code 11, Code 128, Code 32, Code 39, Code 93, GS1 DataBar, Hong Kong 2 of 5, IATA 2 of 5, Interleaved 2 of 5, Matrix 2 of 5, MSI Plessey, NEC 2 of 5, Pharmacode, Plessey, Straight 2 of 5, Telepen, UPC / EAN / JAN, Trioptic
Méthode de transfert de donnée Bluetooth™ 5 Low Energy (class II)
Hauteur (métrique) 198 mm
Hauteur (impériale) 7.79 in.
Largeur (métrique) 86 mm
Largeur (impériale) 3.39 in.
Profondeur (métrique) 152 mm
Profondeur (impériale) 5.98 in.
Poids (métrique) 0.53 kg
Temps de charge Charging over cradle (0 to 100 %) 4 hr.
Poids (impériale) 1.168 lb.
Certifications/Conformité CE, UKCA
À utiliser avec (application) Asset tracking, Barcode Labelling, Circuit Board Labelling, General Identification
Vision du champ High Density Field: 30° Horizontal by 20° Vertical; Wide Field: 50° Horizontal by 33.50° Vertical
Humidité 5% to 95% (non-condensing)
Comprend Lecteur de codes-barres V4500
Garantie 3 Years
Type Barcode Reader
Type de connexion RS-232, USB
Alimentation électrique Rechargeable Battery
Résolution 1280 x 960
Alternate Part Number BR-V4500
Show More Show Less
Has lithium-ion battery - must ship ground