Learn More
X50 Tube Premium K2E E.D.T.A. K2, 6 ml, 13x100,Vio / No, Etq.P
Marque: Greiner Bio One 456023
X50 Tube Premium K2E E.D.T.A. K2, 6 ml, 13x100,Vio / No, Etq.P
Code nomenclature Nacres: NB.11
Informations supplémentaires : Poids : 0.66000kg
Description
- Inner wall is coated with spray-dried K2E K2EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.