missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The VAM1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Lignée cellulaire | The lysate was created in HEK293T cells, using plasmid ID RC25667 and based on accession number NM_16447. The protein contains a C-terminal DDK tag. |
| À utiliser avec (application) | Western Blot |
| Quantité | 0,1 mg |
| Tampon de stockage | RIPA buffer |
| Pureté | Protein |
| Spécificité du test | Homo sapiens membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6) (MPP6), mRNA. |
| Contenu et stockage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Usage exclusivement réservé à la recherche
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?