Learn More
Tocris Bioscience™ VH 298
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
VH 298 is a high-affinity inhibitor of E3 ubiquitin ligase VHL (Kd = 80-90 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. Results in time- and concentration-dependent accumulation of hydroxylated HIF-α, and upregulates mRNA and protein levels of HIF target genes, with similar transcriptional profile to hypoxia. Cell permeable.
Negative control cis VH 298 also available.
Spécification
Spécification
| Quantité | 50 mg |
| Cible | Ubiquitin Ligase (E3) |
| Formule moléculaire | C27H33N5O4S |
| Poids de la formule | 523.65g/mol |
| Type de produit | Inhibiteur |
Usage exclusivement réservé à la recherche