Learn More
Ixazomib, MedChemExpress
Code produit. 16450337
Encompass_Preferred
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 16450337
Marque: Medchem Express HY10453
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
MedChemExpress Ixazomib (MLN2238) is a selective, potent, and reversible proteasome inhibitor, which inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC50 of 3.4 nM (Ki of 0.93 nM).
Ixazomib (MLN2238) is a selective, potent, and reversible proteasome inhibitor, which inhibits the chymotrypsin-like proteolytic (β5) site of the 20S proteasome with an IC50 of 3.4 nM (Ki of 0.93 nM).
Specifications
Nom chimique ou matériau Ixazomib
Forme physique Solid
CAS 1072833-77-2
Couleur Blanc
Quantité 50 mg
Notes de qualité de la pureté Research
Formule moléculaire C14H19BCl2N2O4
À utiliser avec (application) Cancer-Kinase/protease
Synonyme MLN2238
Informations sur la solubilité DMSO : 62.5 mg/mL (173.12 mM; Need ultrasonic)
SMILES OB([C@@H](NC(CNC(C1=CC(Cl)=CC=C1Cl)=O)=O)CC(C)C)O
Poids moléculaire (g/mol) 361.03
Poids de la formule 361.03
Pourcentage de pureté 96.67%
Qualité Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.