Learn More
Novus Biologicals™ Recombinant Human ATBF1/ZFHX3 GST (N-Term) Protein
Tous les produits Bio Techne ProduitsDescription
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spécification
Spécification
| À utiliser avec (application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formule | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Identification génétique (Entrez) | 463 |
| Poids moléculaire | M.W. (Theoretical): 36.74 kDa |
| Quantité | 10 μg |
| Conditions de stockage | Store at -80C. Avoid freeze-thaw cycles. |
| Alias de gène | Alpha-fetoprotein enhancer-binding protein, AT motif-binding factor, AT motif-binding factor 1, ATBF1alpha-fetoprotein enhancer binding protein, AT-binding transcription factor 1ATBT, ZFH-3, zinc finger homeobox 3, zinc finger homeobox protein 3, Zinc finger homeodomain protein 3, ZNF927 |
| Symbole de gène(s) | ZFHX3 |
| Catégorie de recherche | Prostate Cancer |
| Pureté ou qualité | >80% by SDS-PAGE and Coomassie blue staining |
| Afficher plus |