Learn More
Novus Biologicals™ Recombinant Human Kua-UEV GST (N-Term) Protein
Tous les produits Bio Techne ProduitsDescription
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spécification
Spécification
| À utiliser avec (application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formule | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Identification génétique (Entrez) | 387522 |
| Poids moléculaire | MolecularWeight-theroretical: 33.55 kDa |
| Quantité | 25 μg |
| Conditions de stockage | Store at -80°C. Avoid freeze/thaw cycles. |
| Alias de gène | CROC1, CROC-1, CROC1B, CROC-1 B, KUA-UEV, TMEM189-UBE2V1 fusion protein, TMEM189-UBE2V1 readthrough, TMEM189-UBE2V1 readthrough transcript, TRAF6-regulated IKK activator 1 beta Uev1A, transmembrane protein 189-ubiquitin-conjugating enzyme E2 variant 1 read-through, UBE2V, UEV1, UEV-1 |
| Symbole de gène(s) | TMEM189-UBE2V1 |
| Pureté ou qualité | >80% by SDS-PAGE and Coomassie blue staining |
| Protéine | Kua-UEV |