missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-55796
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Vinculin Polyclonal specifically detects Vinculin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Spécification
| Vinculin | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Knockdown Validated | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| VCL | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SALTSKLADLRRQGKGDSPEARALAKQVATALQNLQTKTNRAVANSRPAKAAVHLEGKIEQAQRWIDNPTVDDRGVGQAAIRGLVAEGHRLANVMMGP | |
| 100 μL | |
| Cellular Markers, Cytoskeleton Markers | |
| 7414 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu