missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation XBridge BEH Shield RP18 XP Column
Click to view available options
Longueur (métrique):
30 mm
50 mm
75 mm
100 mm
150 mm
Diamètre (métrique):
2,1 mm
3 mm
4,6 mm
Quantité:
1 de chaque
3/paquet
Conditionnement:
1 pièce
3 pièces
Description
- The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
- Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Specifications
Specifications
| Diamètre (métrique) | 3 mm |
| Longueur (métrique) | 50 mm |
| Pression max. | 18000 psi (1240 Bar) |
| Forme des particules | Sphérique |
| Granulométrie | 2.5 μm |
| Porosité | 130 Å |
| Quantité | 1 de chaque |
| Type d’USP | L1 |
| “End-capped” | Oui |
| Type | eXtended Performance (XP) Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction