Learn More
Waters Corporation XTerra Shield RP18 Column
Code produit. 13202620
Encompass
Click to view available options
Longueur (métrique):
20 mm
30 mm
50 mm
100 mm
150 mm
250 mm
Diamètre (métrique):
2,1 mm
3 mm
3,9 mm
4,6 mm
Granulométrie:
3,5 μm
5 μm
missing translation for 'unitSize'
1 pièce
This item is not returnable. View return policy
Product Code. 13202620
missing translation for 'mfr': Waters Corporation 186000468
This item is not returnable. View return policy
XTerra RP18 columns are general purpose, hybrid-based, reversed-phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds.
  • These first generation columns provide superior pH stability compared to a silica-based column.
Specifications
Diamètre (métrique) 3 mm
Longueur (métrique) 100 mm
Pression max. 6000 psi (415 Bar)
Forme des particules Sphérique
Granulométrie 5 μm
Porosité 125 Å
Quantité 1/Ea.
Type d’USP L1
“End-capped” Oui
Type Colonne
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.