CAS RN 1642288-47-8
CAS RN 1642288-47-8
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
Mavacamten, MedChemExpress
MedChemExpress Mavacamten (MYK461) is an orally active modulator of cardiac myosin, with IC50s of 490, 711 nM for bovine cardiac and human cardiac, respectively.