CAS RN 3093-35-4
CAS RN 3093-35-4
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
Promotions disponibles
Thermo Scientific Chemicals Halcinonide
Thermo Scientific Chemicals Halcinonide
2
Halcinonide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Halcinonide, MedChemExpress
MedChemExpress Halcinonide (SQ-18566) is a high potency corticosteroid used topically in the treatment of certain skin conditions.