CAS RN 3868-31-3
CAS RN 3868-31-3
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
8-Hydroxyguanosine, MedChemExpress
MedChemExpress 8-Hydroxyguanosine is a systematic marker of oxidative stress and a marker of hydroxyl radical damage to RNA.