CAS RN 666260-75-9
CAS RN 666260-75-9
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
GW842166X, MedChemExpress
MedChemExpress GW842166X is a potent and selective cannabinoid receptor 2 (CB2) agonist with IC50 values of 63 and 91 nM for human and rat CB2, respectively.