missing translation for 'onlineSavingsMsg'
Learn More

Acide 1-phényl-1H-pyrazole-4-carboxylique, 99 %, Thermo Scientific Chemicals

Catalog Number p-7033236
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1134-50-5
C10H8N2O2
188.186
MFCD00514518
ZROILLPDIUNLSE-UHFFFAOYSA-N
1-phenyl-1h-pyrazole-4-carboxylic acid, pyrazole-4-carboxylic acid, 1-phenyl, 1h-pyrazole-4-carboxylic acid, 1-phenyl, 1-phenylpyrazole-4-carboxylate, 1-phenyl-1h-pyrazol-4-carboxylic acid, pyrazole, 1, 1-phenyl-1h-pyrazole-4-carboxylicacid, akos pao-0321, acmc-1c9e6
121026
Acide 1-phénylpyrazole-4-carboxylique
C1=CC=C(C=C1)N2C=C(C=N2)C(=O)O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1134-50-5
C10H8N2O2
188.186
MFCD00514518
ZROILLPDIUNLSE-UHFFFAOYSA-N
1-phenyl-1h-pyrazole-4-carboxylic acid, pyrazole-4-carboxylic acid, 1-phenyl, 1h-pyrazole-4-carboxylic acid, 1-phenyl, 1-phenylpyrazole-4-carboxylate, 1-phenyl-1h-pyrazol-4-carboxylic acid, pyrazole, 1, 1-phenyl-1h-pyrazole-4-carboxylicacid, akos pao-0321, acmc-1c9e6
121026
Acide 1-phénylpyrazole-4-carboxylique
C1=CC=C(C=C1)N2C=C(C=N2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 1-Phenyl-1H-pyrazole-4-carboxylic acid
Quantité 250 mg
Poids de la formule 188.19
Pourcentage de pureté 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'