missing translation for 'onlineSavingsMsg'
Learn More

Methandienone, Lipomed

Code produit. 30459151
Click to view available options
Quantité:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30459151

Brand: Lipomed LPMTES124910

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Methandienone
TRUSTED_SUSTAINABILITY

Specifications

Nom chimique ou matériau Methandienone
CAS 72-63-9
Noms d’analytes ou de composants Methandienone
Conditions d’expédition Room Temperature
Formule moléculaire C20 H28 O2
Synonyme Androsta-1,4-dien-3-one, 17-hydroxy-17-methyl-, (17β)-, Androsta-1,4-dien-3-one, 17β-hydroxy-17-methyl- (8 CI), (17β)-17-Hydroxy-17-methylandrosta-1,4-dien-3-one, 1,2-Dehydro-17-methyltestosterone, 1,4-Androstadien-17α-methyl-17β-ol-3-one, 1-Dehydro-17α-methyltestosterone, 1-Dehydromethyltestosterone, 17-Methylandrosta-1,4-dien-17β-ol-3-one, 17α-Methyl-1-dehydrotestosterone, 17α-Methyl-17β-hydroxyandrost-1,4-dien-3-one, 17α-Methyl-17β-hydroxyandrosta-1,4-dien-3-one, 17α-Methylandrosta-1,4-dien-17β-ol-3-one, 17β-Hydroxy-17-methylandrosta-1,4-dien-3-one, 17β-Hydroxy-17α-methyl-1,4-androstadiene-3-one, 17β-Hydroxy-17α-methylandrosta-1,4-dien-3-one, Abirol, Anabolin, Ciba 17309-Ba, Compound 17309, Crein, Danabol, Dehydromethyltestosterone, Dianabol, Dianabole, Geabol, MA, Metanabol, Metandienon, Metandienone, Metandienonum, Metandrostenolon, Metandrostenolone, Metastenol, Methandienone, Methandon, Methandrolone, Methandrostenolone, Methylboldenone, NSC 51180, NSC-42722, Nabolin, Naposim, Nerobol, Nerobolettes, Protobolin, Stenolon, Stenolone, Sterolon, Δ1-17α-Methyltestosterone
Formule InChI InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES C[C@]1(O)CC[C@H]2 [C@@H]3 CCC4=CC(=O)C=C[C@]4(C)[C@H]3 CC[C@]12 C
Nom IUPAC (8 R,9 S,10 R,13 S,14 S,17 S)-17-hydroxy-10,13,17-trimethyl-7,8,9,11,12,14,15,16-octahydro-6 H-cyclopenta[a]phenanthren-3-one
Industry Type Pharmaceutical
Poids moléculaire (g/mol) 300.44
Quantité 10 mg
Poids de la formule 300.2089 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.