Learn More
2,5-Dimethylpyrrole-3-acide carboxylique, 97 %, Thermo Scientific Chemicals
Catalog Number p-7025246
Click to view available options
Quantité:
1 g
5 g
25 g
57338-76-8
C7H9NO2
139.154
MFCD00203874
VDVWTJFVFQVCFN-UHFFFAOYSA-N
2,5-dimethylpyrrole-3-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl, 2,5-dimethyl-4-carboxypyrrole, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl
292957
2,5-diméthyle-1H-pyrrole-3-acide carboxylique
CC1=CC(=C(N1)C)C(=O)O
57338-76-8
C7H9NO2
139.154
MFCD00203874
VDVWTJFVFQVCFN-UHFFFAOYSA-N
2,5-dimethylpyrrole-3-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl, 2,5-dimethyl-4-carboxypyrrole, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl
292957
2,5-diméthyle-1H-pyrrole-3-acide carboxylique
CC1=CC(=C(N1)C)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 2,5-Dimethylpyrrole-3-carboxylic acid
Point de fusion 199°C to 201°C (decomposition)
Quantité 1 g
Poids de la formule 139.16
Pourcentage de pureté 97%