missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCML2 is a putative component of a Polycomb group (PcG) multiprotein complex involved in represing transcription of homeotic genes during development. It mediates chromatin remodeling and DNA dependent negative regulation of transcription by binding to methylated lysine residues in histone tails.
Spécification
Spécification
| Antigène | SCML2 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Expression | SCML2 |
| Numéro d’ordre du gène | Q9UQR0 |
| Alias de gène | SCML2 |
| Symboles de gène(s) | SCML2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?