missing translation for 'onlineSavingsMsg'
Learn More

Acide 2-méthyl-4-(trifluorométhoxy)cinnamique, 97 %, Thermo Scientific™

Catalog Number 15425867
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1588508-08-0
C11H9F3O3
246.19
MFCD18399792
JYTHJDZEVRTJAK-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy cinnamic acid, 2e-3-2-methyl-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-2-methyl-4-trifluoromethoxy phenyl acrylic acid
86277666
Acide (E)-3-[2-méthyl-4-(trifluorométhoxy)phényl]prop-2-énoïque
CC1=C(C=CC(O)=O)C=CC(OC(F)(F)F)=C1
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1588508-08-0
C11H9F3O3
246.19
MFCD18399792
JYTHJDZEVRTJAK-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy cinnamic acid, 2e-3-2-methyl-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-2-methyl-4-trifluoromethoxy phenyl acrylic acid
86277666
Acide (E)-3-[2-méthyl-4-(trifluorométhoxy)phényl]prop-2-énoïque
CC1=C(C=CC(O)=O)C=CC(OC(F)(F)F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 2-Methyl-4-(trifluoromethoxy)cinnamic acid
Quantité 1 g
Poids de la formule 246.18
Pourcentage de pureté 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'