missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 4-1BB Ligand Recombinant Rat Monoclonal Antibody (AT113-2)

Code produit 17951361
Change view
Click to view available options
Quantité:
200 μg
Conditionnement:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17951361 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17951361 Fournisseur Invitrogen™ Code fournisseur MA548152

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rat Recombinant Monoclonal Antibody

Specificity: This antibody is specific for 4-1BB ligand (also known as CD137 ligand), which is a member of the TNF superfamily and is a type II membrane protein.

CD137 ligand (CD137L) is a type II membrane protein and part of the TNF superfamily. CD137L is a co-stimulatory molecule that is expressed on antigen presenting cells (DCs, monocytes/macrophages, B cells) and is upregulated upon activation. Its receptor, CD137/4-1BB, is found on a variety of cells, including inflamed endothelial cells, where its expression enhances extravasation of CD137L expressing monocytes. Activated T cells also express CD137, and engagement with CD137L enhances T cell proliferation, IL-2 secretion, survival, and cytotoxic activity. CD137L is also expressed on a majority of B-cell lymphomas, including mantle cell lymphoma, follicular lymphoma, and diffuse large B-cell lymphoma (but it is lacking on Hodgkin lymphoma and T-cell lymphoma). CD137L is also expressed in carcinoma cell lines, and is thought to be involved in T cell-tumor cell interactions.
TRUSTED_SUSTAINABILITY

Spécification

Antigène 4-1BB Ligand
Applications Flow Cytometry, Inhibition Assays, Western Blot
Classification Recombinant Monoclonal
Clone AT113-2
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.02% ProClin 300
Expression TNFSF9
Numéro d’ordre du gène P41274
Alias de gène 4 1 BB Ligand; 4 1 BBL; 41BB Ligand; 4-1BB ligand; 41BBL; 4-1BBL; 4-1BB-L; AI848817; CD 137 Ligand; CD 137L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1BB-L; Ly63l; receptor 4-1BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9
Symboles de gène(s) TNFSF9
Espèces hôtes Rat
Immunogène This antibody was raised by immunising LOU rats with soluble recombinant 4-1BBL:Fc fusion protein.
Méthode de purification Protein A
Quantité 200 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 21950
Espèces cibles Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.