Learn More
Ester pinacolique d’acide boronique de 4-(Phénoxyméthyl)benzène, 95 %, Thermo Scientific™
Catalog Number p-7034596
Click to view available options
Quantité:
1 g
5 g
946409-21-8
C19H23BO3
310.20
MFCD18827485
YLEMWKPHWXIWOL-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-4-phenoxymethyl-phenyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-4-phenoxymethyl phenyl-1,3,2-dioxaborolane, amtb380, 2-4-phenoxymethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
57655492
4,4,5,5-tétraméthyl-2-[4-phénoxyméthyl)phényl]-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=C(COC2=CC=CC=C2)C=C1
946409-21-8
C19H23BO3
310.20
MFCD18827485
YLEMWKPHWXIWOL-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-4-phenoxymethyl-phenyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-4-phenoxymethyl phenyl-1,3,2-dioxaborolane, amtb380, 2-4-phenoxymethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
57655492
4,4,5,5-tétraméthyl-2-[4-phénoxyméthyl)phényl]-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1=CC=C(COC2=CC=CC=C2)C=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4-(Phenoxymethyl)benzeneboronic acid pinacol ester
Quantité 1 g
Poids de la formule 310.2
Pourcentage de pureté 95%