missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-iodo-1-méthyl-1H-pyrazole, 97 %, Thermo Scientific™
71.95€ - 208.00€
Identifiants chimiques
CAS | 34091-51-5 |
---|---|
Formule moléculaire | C4H5IN2 |
Poids moléculaire (g/mol) | 208.00 |
Numéro MDL | MFCD11109327 |
Clé InChI | RJYWUQWCLZYCTI-UHFFFAOYSA-N |
CID PubChem | 13334065 |
Nom IUPAC | 5-iodo-1-methyl-1H-pyrazole |
SMILES | CN1N=CC=C1I |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
34091-51-5 | |
208.00 | |
RJYWUQWCLZYCTI-UHFFFAOYSA-N | |
5-iodo-1-methyl-1H-pyrazole |
C4H5IN2 | |
MFCD11109327 | |
13334065 | |
CN1N=CC=C1I |
Specifications
5-Iodo-1-methyl-1H-pyrazole | |
78°C to 81°C | |
MFCD11109327 | |
5-iodo-1-methyl-1h-pyrazole, 1-methyl-5-iodopyrazole, 1h-pyrazole,5-iodo-1-methyl, 1h-pyrazole, 5-iodo-1-methyl, 1-methyl-5-iodo-1h-pyrazole | |
CN1N=CC=C1I | |
208.00 | |
208 |
34091-51-5 | |
C4H5IN2 | |
Light and moisture sensitive | |
RJYWUQWCLZYCTI-UHFFFAOYSA-N | |
5-iodo-1-methyl-1H-pyrazole | |
13334065 | |
97% |
Safety and Handling
Déclaration GHS H
H318-H335-H315
Provoque de graves lésions des yeux.
Peut irriter les voies respiratoires.
Provoque une irritation cutanée.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : Non
Recommended Storage : 2° à 8°C