Learn More
Hamilton™ 500 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Marque: Hamilton™ CAL80801
Informations supplémentaires : Poids : 0.03100kg
Description
- This series of syringes are ideal for dispensing volumes up to 500 μL.
- A variety of different terminations and needle options are available in this series.
Spécification
Autoclavable | |
No Needle Included | |
0.305 in. | |
7.75 mm | |
Gas Tight Syringe | |
115°C | |
1750 RN Syringe | |
1 | |
No Needle Included | |
Removable |
Stainless Steel | |
0.128 in. | |
3.26 mm | |
No Needle Included | |
239°F | |
50°F | |
10°C | |
500 μL | |
No Needle Included |