Learn More
Hamilton™ 500 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Marque:  Hamilton™ CAL81256
Informations supplémentaires : Poids : 0.05800kg
Code produit. 17824282
  • 424.00€ / 1 pièce
Expédition estimée: 27-06-2024
Description
  • This series of syringes are ideal for dispensing volumes up to 500 μL.
  • A variety of different terminations and needle options are available in this series.
Spécification
Spécification
Autoclavable
22 G
0.0161 in.
0.0283 in.
3.26 mm
0.72 mm
Gas Tight Syringe
115°C
1750 Syringe
1
51 mm
Removable
Borosilicate Glass
0.128 in.
0.41 mm
0.305 in.
7.75 mm
2 in.
239°F
50°F
10°C
500 μL
Sharp, Bevel, Curved, Non-coring