Learn More
Univet™ 506UP Clear Safety Glasses
Click to view available options
:
506UP Clear Safety Glasses; Material: Polycarbonate
Description
- Lens available with Vanguard PLUS coating
- Optimal comfort and fit thanks to the soft adjustable nose pad
- Soft terminal tips to eliminate local pressure
Spécification
Spécification
| Forme | Rectangular |
| Revêtement de la lentille | Anti-fog, Scratch-resistant |
| Teinte de la lentille | None |
| Type de produit | Safety Glasses |
| Matériau | Polycarbonate |
| Jetable | Reusable |
| Lens Material | Polycarbonate |
| Matériau de la monture | Polycarbonate |
| Protection UV | UV380 |
| Frame Type | Adjustable Temples |
| Afficher plus |