Learn More
Phenyramidol Salicylate, Mikromol™
Code produit. 30052627
Click to view available options
Quantité:
50 mg
Conditionnement:
50mg
Code produit. 30052627
Marque: Mikromol MM3189.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Phenyramidol Salicylate
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau Phenyramidol Salicylate
CAS 1235-07-0
Noms d’analytes ou de composants Phenyramidol Salicylate
Conditions d’expédition Room Temperature
Formule moléculaire C13 H14 N2 O . C7 H6 O3
Certifications/Conformité ISO 17025
Synonyme Analexin syrup, Phenyramidol salicylate, Benzoic acid, 2-hydroxy-, compd. with alpha-[(2-pyridinylamino)methyl]benzenemethanol (1:1), Salicylic acid, compd. with alpha-[(2-pyridylamino)methyl]benzyl alcohol (1:1) (8CI), Salicylic acid, compd. with alpha-[(2-pyridylamino)methyl]benzyl alcohol (7CI), Benzenemethanol, alpha-[(2-pyridinylamino)methyl]-, mono(2-hydroxybenzoate) (salt) (9CI), Benzyl alcohol, alpha-[(2-pyridylamino)methyl]-, monosalicylate (salt)
Formule InChI InChI=1S/C13H14N2O.C7H6O3/c16-12(11-6-2-1-3-7-11)10-15-13-8-4-5-9-14-13;8-6-4-2-1-3-5(6)7(9)10/h1-9,12,16H,10H2,(H,14,15);1-4,8H,(H,9,10)
SMILES OC(CNc1ccccn1)c2ccccc2.OC(=O)c3ccccc3O
Nom IUPAC 2-hydroxybenzoic acid;1-phenyl-2-(pyridin-2-ylamino)ethanol
Poids moléculaire (g/mol) 352.38
Quantité 50 mg
Poids de la formule 352.1423 g/mol