Learn More
Uvex™ 9172 Super G Safety Glasses
Tous les produits Uvex Produits
Click to view available options
:
Supravision Sapphire
Description
- The lightest safety glasses in the world weighing only 18 grams with full safety, modern design and an innovative temple concept
- Supravision excellence coating is anti-fog on the inside, and extremely scratch-resistant and chemical-resistant on the outside
- Supravision plus coating is anti-fog and scratch-resistant on both sides
- Supravision sapphire coating is extremely scratch-resistant and chemical-resistant on both sides
- Aerodynamically optimized and inclined lens geometry
- Soft temple ends for maximum wearing comfort
- Easy lens replacement
Spécification
Spécification
| Type de lentille | Supravision Sapphire |
| Certifications / conformité (Europe) | EN 166-2001, EN 170-2002 |
| Teinte de la lentille | Clear |
| Couleur de la monture | Gris |
| Jetable | Reusable |
| Lens Material | Polycarbonate |
| Protection UV | UV 400 |
| Autoclavable | Non autoclavable |