missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCB10 Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The function of this mitochondrial protein is unknown.
Spécification
Spécification
| Antigène | ABCB10 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Expression | ABCB10 |
| Numéro d’ordre du gène | Q9JI39, Q9NRK6 |
| Alias de gène | ABC transporter 10 protein; ABCB10; Abcb12; Abc-me; ABC-me protein; ABC-mitochondrial erythroid protein; ATP binding cassette subfamily B member 10; ATP-binding cassette sub-family B member 10, mitochondrial; ATP-binding cassette transporter 10; ATP-binding cassette, subfamily B (MDR/TAP), member 10; ATP-binding cassette, sub-family B (MDR/TAP), member 10; EST20237; M-ABC2; mitochondrial ATP-binding cassette 2; MTABC2; si:dkey-162b3.3 |
| Symboles de gène(s) | ABCB10 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?