Learn More
AKAP8L Rabbit anti-Human, Polyclonal, Proteintech
Code produit 16886263 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16886263 20 μL 20µL
16876263 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16886263 Fournisseur Proteintech Code fournisseur 101031AP20UL
Rabbit Polyclonal Antibody
Could play a role in constitutive transport element (CTE)-mediated gene expression. Does not seem to be implicated in the binding of regulatory subunit II of PKA. May be involved in nuclear envelope breakdown and chromatin condensation. May regulate the initiation phase of DNA replication when associated with TMPO-beta.
Spécification
Antigène AKAP8L
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Expression AKAP8L
Numéro d’ordre du gène Q9ULX6
Alias de gène A kinase anchor protein 8 like, AKAP8 like protein, AKAP8L, HA95, HAP95, Helicase A binding protein 95, Homologous to AKAP95 protein, NAKAP, NAKAP95, Neighbor of AKAP95
Symboles de gène(s) AKAP8L
Espèces hôtes Rabbit
Immunogène AKAP8L Fusion Protein Ag0145
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 26993
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG