Learn More
Tube AL-23, ouvert aux deux extrémités, dia. ext. : 16 mm, dia. int. de : 12mm, Thermo Scientific™
Marque: Thermo Scientific Alfa Aesar 031568.EH
Code nomenclature Nacres: NA.23
Informations supplémentaires : Poids : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Tube | |
Both ends open | |
600 mm |